MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Salvianolic acid B protects against acute ethanol-induced liver injury through SIRT1-mediated deacetylation of p53 in rats.
Salvianolic acid B protects against acute ethanol-induced liver injury through SIRT1-mediated deacetylation of p53 in rats.
Toxicology letters (2014-04-29)
Mingzhu Li, Yang Lu, Yan Hu, Xiaohan Zhai, Wei Xu, Huirong Jing, Xiaofeng Tian, Yuan Lin, Dongyan Gao, Jihong Yao
ABSTRACT
Salvianolic acid B (SalB) is isolated from the traditional Chinese medical herb salvia miltiorrhiza. It has many biological and pharmaceutical activities. This study aimed to investigate the effect of SalB on acute ethanol-induced hepatic injury in rats and to explore the role of SIRT1 in this process. The results showed that pretreatment with SalB significantly reduced ethanol-induced elevation in aminotransferase activities, decreased hepatotoxic cytokine levels such as Interleukin-6 (IL-6), and increased the antioxidant enzyme activity. Moreover, SalB pretreatment reversed the increase in NF-κB, cleaved caspase-3 and decrease in B-cell lymphoma-extra large (Bcl-xL) caused by ethanol exposure. Importantly, SalB pretreatment significantly increased the expression of SIRT1, a NAD(+)-dependent deacetylase, whereas the increase in SIRT1 was accompanied by decreased acetyl-p53 expression. In HepG2 cells, SalB pretreatment increased SIRT1 expression in a time and dose-dependent manner and such an increase was abrogated by siRNA knockdown of SIRT1. Additionally, inhibition of SIRT1 significantly increased the acetylation of p53, and blocked SalB-induced acetylation of p53 down-regulation. Collectively, this study indicated that SalB can alleviate acute ethanol-induced hepatocyte apoptosis through SIRT1-mediated deacetylation of p53 pathway.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Reagent Alcohol, denatured, suitable for HPLC
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, denatured, denatured, reagent grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC