MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Novel biosensors based on optimized glycine oxidase.
The FEBS journal (2014-06-14)
Elena Rosini, Luciano Piubelli, Gianluca Molla, Luca Frattini, Mattia Valentino, Antonio Varriale, Sabato D'Auria, Loredano Pollegioni
ABSTRACT
Glycine is involved in several physiological functions, e.g. as a neurotransmitter in the central nervous system, and sarcosine has been identified as a differential metabolite greatly enhanced during prostate cancer progression and metastasis. Glycine oxidase from Bacillus subtilis (GO) was engineered with the final aim of producing specific analytical systems to detect these small achiral amino acids. Based on in silico analysis, site-saturation mutagenesis was independently performed at 11 positions: a total of 16 single-point GO variants were analyzed. Significantly improved kinetic parameters were observed on glycine for the A54R, H244K-N-Q-R, Y246W and M261R variants. The introduction of multiple mutations then identified the H244K/M261R variant showing a 5.4-fold increase in maximal activity on glycine. With sarcosine as substrate, a number of single-point variants showed increased maximal activity and/or affinity: the kinetic efficiency was increased 6-fold for the M49L variant. Two GO variants with a high substrate specificity ratio for glycine (versus sarcosine, i.e. H244K GO) or for sarcosine (versus glycine, i.e. M49L GO) combined with high substrate affinity were used to set up a simple fluorescence-based biosensor. This optical sensing assay represents a novel, inexpensive and fast tool to assay glycine or sarcosine concentrations in biological samples (detection limit ≤ 0.5 μm).
Product Description
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sarcosine, BioXtra
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)