MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A mutation in the DNA polymerase accessory factor of herpes simplex virus 1 restores viral DNA replication in the presence of raltegravir.
A mutation in the DNA polymerase accessory factor of herpes simplex virus 1 restores viral DNA replication in the presence of raltegravir.
Journal of virology (2014-07-11)
Bin Zhou, Kui Yang, Elizabeth Wills, Liang Tang, Joel D Baines
ABSTRACT
Previous reports showed that raltegravir, a recently approved antiviral compound that targets HIV integrase, can inhibit the nuclease function of human cytomegalovirus (HCMV terminase) in vitro. In this study, subtoxic levels of raltegravir were shown to inhibit the replication of four different herpesviruses, herpes simplex virus 1 (HSV-1), HSV-2, HCMV, and mouse cytomegalovirus, by 30- to 700-fold, depending on the dose and the virus tested. Southern blotting and quantitative PCR revealed that raltegravir inhibits DNA replication of HSV-1 rather than cleavage of viral DNA. A raltegravir-resistant HSV-1 mutant was generated by repeated passage in the presence of 200 μM raltegravir. The genomic sequence of the resistant virus, designated clone 7, contained mutations in 16 open reading frames. Of these, the mutations F198S in unique long region 15 (UL15; encoding the large terminase subunit), A374V in UL32 (required for DNA cleavage and packaging), V296I in UL42 (encoding the DNA polymerase accessory factor), and A224S in UL54 (encoding ICP27, an important transcriptional regulator) were introduced independently into the wild-type HSV-1(F) genome, and the recombinant viruses were tested for raltegravir resistance. Viruses bearing both the UL15 and UL32 mutations inserted within the genome of the UL42 mutant were also tested. While the UL15, UL32, and UL54 mutant viruses were fully susceptible to raltegravir, any virus bearing the UL42 mutation was as resistant to raltegravir as clone 7. Overall, these results suggest that raltegravir may be a valuable therapeutic agent against herpesviruses and the antiviral activity targets the DNA polymerase accessory factor rather than the nuclease activity of the terminase. This paper shows that raltegravir, the antiretrovirus drug targeting integrase, is effective against various herpesviruses. Drug resistance mapped to the herpesvirus DNA polymerase accessory factor, which was an unexpected finding.
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
DNA Polymerase I, Klenow Fragment from Escherichia coli, buffered aqueous glycerol solution
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2