MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The cytotoxic and genotoxic effects of metalaxy-M on earthworms (Eisenia fetida).
The cytotoxic and genotoxic effects of metalaxy-M on earthworms (Eisenia fetida).
Environmental toxicology and chemistry (2014-07-22)
Tong Liu, Lusheng Zhu, Yingnan Han, Jinhua Wang, Jun Wang, Yan Zhao
ABSTRACT
As the main optical isomer of metalaxyl, metalaxyl-M has been widely used worldwide in recent years because of its notable effect on the prevention and control of crop diseases. Together with the toxicity and degradation of metalaxyl-M, the chemical has attracted the attention of researchers. The present study examined the toxic effects of metalaxyl-M on earthworms at 0 mg kg(-1) , 0.1 mg kg(-1) , 1 mg kg(-1) , and 3 mg kg(-1) on days 7, 14, 21 and 28 after exposure. The results showed that metalaxyl-M could cause an obvious increase in the production of reactive oxygen species (ROS) when the concentration was higher than 0.1 mg kg(-1) , which led to lipid peroxidation in earthworms. Metalaxyl-M can induce DNA damage in earthworms, and the level of DNA damage markedly increased with increasing the concentration of metalaxyl-M. Metalaxyl-M also has a serious influence on the activities of antioxidant enzymes, which results in irreversible oxidative damage in cells. The changes of these indicators all indicated that metalaxyl-M may cause cytotoxic and genotoxic effects on earthworms.
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Supelco
Metalaxyl, PESTANAL®, analytical standard
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, ≥99%
Supelco
Metalaxyl-M, PESTANAL®, analytical standard
Alanine, European Pharmacopoeia (EP) Reference Standard