MilliporeSigma
  • Inhibition effect of glyphosate on the acute and subacute toxicity of cadmium to earthworm Eisenia fetida.
Inhibition effect of glyphosate on the acute and subacute toxicity of cadmium to earthworm Eisenia fetida.
Environmental toxicology and chemistry (2014-07-22)
Chui-Fan Zhou, Yu-Jun Wang, Rui-Juan Sun, Cun Liu, Guang-Ping Fan, Wen-Xiu Qin, Cheng-Cheng Li, Dong-Mei Zhou
ABSTRACT
The acute and subacute toxicities of cadmium (Cd) to earthworm Eisenia fetida in the presence and absence of glyphosate were studied. Although Cd is highly toxic to E. fetida, the presence of glyphosate markedly reduced the acute toxicity of Cd to earthworm; both the mortality rate of the earthworms and the accumulation of Cd decreased with the increase of the glyphosate/Cd molar ratio. The subcellular distribution of Cd in E. fetida tissues showed that internal Cd was dominant in the intact cells fraction and the heat-stable proteins fraction. The presence of glyphosate reduced the concentration of Cd in all fractions, especially the intact cells. During a longer period of exposure, the weight loss of earthworm and the total Cd absorption was alleviated by glyphosate. Thus, the herbicide glyphosate can reduce the toxicity and bioavailability of Cd in the soil ecosystems at both short- and long-term exposures.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%
Cadmium, wire reel, 0.2m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 1m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%