MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Treatment-related fatigue with sorafenib, sunitinib and pazopanib in patients with advanced solid tumors: an up-to-date review and meta-analysis of clinical trials.
Treatment-related fatigue with sorafenib, sunitinib and pazopanib in patients with advanced solid tumors: an up-to-date review and meta-analysis of clinical trials.
International journal of cancer (2014-01-15)
Matteo Santoni, Alessandro Conti, Francesco Massari, Giorgio Arnaldi, Roberto Iacovelli, Mimma Rizzo, Ugo De Giorgi, Laura Trementino, Giuseppe Procopio, Giampaolo Tortora, Stefano Cascinu
ABSTRACT
Fatigue is the most common symptom associated with cancer and cancer treatment. We performed an up-to-date meta-analysis to determine the incidence and relative risk (RR) of fatigue in patients (pts) with cancer treated with sorafenib (SO), sunitinib (SU) and pazopanib (PZ). PubMed databases were searched for articles published till August 2013. Eligible studies were selected according to PRISMA statement. Summary incidence, RR and 95% confidence intervals were calculated using random-effects or fixed-effects models based on the heterogeneity of selected studies. Fifteen studies were included in our analysis. A total of 6,996 pts was enrolled: 2,260 had renal cell carcinomas (RCC), 1,691 non-small cell lung cancers, 1,290 breast cancers, 823 hepatocellular carcinomas, 362 soft tissue sarcomas, 304 gastrointestinal solid tumors, 165 neuroendocrine tumors and 101 melanomas. When stratified by drug, SO registered lower incidence and RR of all and high-grade fatigue when compared to SU, whereas the difference between SO and PZ was significant only for all-grade fatigue (p < 0.001). The difference between SU and PZ was significant for high-grade (p < 0.001) but not for all-grade fatigue (p = 0.52). In RCC pts, PZ showed the lower incidence and RR of all and high-grade fatigue. The differences were significant for SU vs. SO (p < 0.001), SU vs. PZ (p < 0.001) and SO vs. PZ (p < 0.001). Treatment with SO, SU and PZ is associated with an increased incidence of fatigue in pts with cancer. Early and appropriate management is required to avoid unnecessary dose reductions and transitory or definitive treatment discontinuations.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®