MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
The effect of cobalt on the human eye.
Documenta ophthalmologica. Advances in ophthalmology (2014-11-09)
Caroline Ann Lim, Jane Khan, Enid Chelva, Riaz Khan, Timothy Unsworth-Smith
ABSTRACT
The effect of elevated serum cobalt on the human visual system has not yet been established. In light of recent reports of visual problems with elevated cobalt in association with hip prostheses, this review examines the literature for evidence regarding the effects of cobalt on human visual function. A systematic literature review was undertaken in July 2013. The electronic databases of PubMed (1955 to July week 1 2013), Cochrane Library and EMBASE were searched. Only human studies or case reports written in English were included. Eight case reports were identified. Five case reports involved patients with metal hip arthroplasties, two cases involved patients exposed to environmental cobalt, and one case involved a patient treated medically for anaemia with cobalt chloride. No human prospective studies were identified. Several case reports showed that high serum cobalt may be associated with both irreversible and reversible visual loss, optic neuropathy and atrophy, electrophysiological evidence of abnormal retinal and retinal pigment epithelium function and fluorescein angiographic evidence of abnormal choroidal perfusion.
Sigma-Aldrich
Cobalt(II) chloride, 97%
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Sigma-Aldrich
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
Cobalt, foil, 4mm disks, thickness 0.01mm, 99.9%
Cobalt, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cobalt, foil, 10mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%