MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • New therapeutic strategy for hepatocellular carcinoma by molecular targeting agents via inhibition of cellular stress defense mechanisms.
New therapeutic strategy for hepatocellular carcinoma by molecular targeting agents via inhibition of cellular stress defense mechanisms.
Journal of UOEH (2014-12-17)
Yuichi Honma, Masaru Harada
ABSTRACT
The prognosis of advanced hepatocellular carcinoma (HCC) has remained very poor.It has recently been reported that the molecular targeting agent sorafenib can improve the prognosis of patients with advanced HCC. However, the detailed mechanisms of sorafenib, especially its direct effects on hepatoma and hepatocyte cells, are poorly understood, making a more detailed investigation about the molecular mechanism of sorafenib necessary. Endoplasmic reticulum (ER) stress is related to the pathophysiology of various liver diseases, including chronic viral hepatitis, alcoholic and nonalcoholic steatohepatitis and HCC. In this regard, our recent data examining the molecular effects of sorafenib focused on the cellular defense mechanisms from ER stress, the unfolded protein response (UPR) and keratin phosphorylation, demonstrated that sorafenib inhibited both important cytoprotective mechanisms, UPR and keratin phosphorylation, and enhances the anti-tumor effect in combination with proteasome inhibitors. This review summarizes the cytoprotective mechanisms from ER stress and our results about the direct effect of sorafenib on the cytoprotective mechanisms.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®