MilliporeSigma
  • Linifanib versus Sorafenib in patients with advanced hepatocellular carcinoma: results of a randomized phase III trial.
Linifanib versus Sorafenib in patients with advanced hepatocellular carcinoma: results of a randomized phase III trial.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2014-12-10)
Calin Cainap, Shukui Qin, Wen-Tsung Huang, Ik Joo Chung, Hongming Pan, Ying Cheng, Masatoshi Kudo, Yoon-Koo Kang, Pei-Jer Chen, Han-Chong Toh, Vera Gorbunova, Ferry A L M Eskens, Jiang Qian, Mark D McKee, Justin L Ricker, Dawn M Carlson, Saied El-Nowiem
ABSTRACT
This open-label phase III trial evaluated efficacy and tolerability of linifanib versus sorafenib in patients with advanced hepatocellular carcinoma (HCC) without prior systemic therapy. Patients were randomly assigned in a 1:1 ratio to linifanib 17.5 mg once daily or sorafenib 400 mg twice daily. Patients were stratified by region (Outside Asia, Japan, and rest of Asia), Eastern Cooperative Oncology Group performance score (ECOG PS; 0 or 1), vascular invasion or extrahepatic spread (yes or no), and hepatitis B virus (HBV) infection (yes or no). The primary end point of the study was overall survival (OS). Secondary end points were time to progression (TTP) and objective response rate (ORR) per RECIST v1.1. We randomly assigned 1,035 patients (median age, 60 years; Asian, 66.6%; ECOG PS 0, 65.2%; HBV, 49.1%; vascular invasion or extrahepatic spread, 70.1%). Median OS was 9.1 months on the linifanib arm (95% CI, 8.1 to 10.2) and 9.8 months on the sorafenib arm (95% CI, 8.3 to 11.0; hazard ratio [HR], 1.046; 95% CI, 0.896 to 1.221). For prespecified stratification subgroups, OS HRs ranged from 0.793 to 1.119 and the 95% CI contained 1.0. Median TTP was 5.4 months on the linifanib arm (95% CI, 4.2 to 5.6) and 4.0 months on the sorafenib arm (95% CI, 2.8 to 4.2; HR, 0.759; 95% CI, 0.643 to 0.895; P = .001). Best response rate was 13.0% on the linifanib arm versus 6.9% on the sorafenib arm. Grade 3/4 adverse events (AEs); serious AEs; and AEs leading to discontinuation, dose interruption, and reduction were more frequent with linifanib (all P < .001). Linifanib and sorafenib had similar OS in advanced HCC. Predefined superiority and noninferiority OS boundaries were not met for linifanib and the study failed to meet the primary end point. TTP and ORR favored linifanib; safety results favored sorafenib.
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Nicotinamide, European Pharmacopoeia (EP) Reference Standard