MilliporeSigma
  • Stem cell factor induces AP-1-dependent mast cell IL-6 production via MAPK kinase 3 activity.
Stem cell factor induces AP-1-dependent mast cell IL-6 production via MAPK kinase 3 activity.
Journal of leukocyte biology (2014-01-24)
Adam J MacNeil, Robert D Junkins, Zhengli Wu, Tong-Jun Lin
ABSTRACT
Mast cells are critical immune effectors abundant in tissues interfacing with the environment and have major roles in allergen-induced inflammation and host responses to infection. SCF is a regulator of mast cell function and growth. However, the critical mechanisms in SCF-directed events remain incompletely defined. Here, we have investigated the role of MKK3 in mast cell SCF signaling-dependent functions by using BMMCs from MKK3-deficient mice. MKK3 was phosphorylated rapidly and persistently following SCF-induced activation and contributed to mast cell proliferation but not survival or migration in response to SCF. Analysis of SCF-induced mast cell mediator secretion demonstrated that IL-6 production is specifically dependent on MKK3 signals, both independently and in concert with IgE. Analysis of SCF-induced signaling showed that sustained p38 phosphorylation was impaired in MKK3-deficient mast cells, where as early JNK and IκBα activation were enhanced. Notably, SCF-inducible expression and activation of c-Jun, a component of the AP-1 transcription factor, was significantly dependent on MKK3. Accordingly, AP-1 DNA-binding activity and interaction with the IL6 gene promoter was markedly impaired in MKK3-deficient mast cells, whereas transcription factors of the Egr family, NF-κB, and NFAT retained near-full activity. These results designate MKK3 as a novel, positive regulator of SCF-induced mast cell proliferation and a critical signaling protein for AP-1-dependent IL-6 production.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture