MilliporeSigma
  • Ostwald's rule of stages governs structural transitions and morphology of dipeptide supramolecular polymers.
Ostwald's rule of stages governs structural transitions and morphology of dipeptide supramolecular polymers.
Nature communications (2014-11-14)
Aviad Levin, Thomas O Mason, Lihi Adler-Abramovich, Alexander K Buell, George Meisl, Celine Galvagnion, Yaron Bram, Samuel A Stratford, Christopher M Dobson, Tuomas P J Knowles, Ehud Gazit
ABSTRACT
The self-assembly of molecular building blocks into nano- and micro-scale supramolecular architectures has opened up new frontiers in polymer science. Such supramolecular species not only possess a rich set of dynamic features as a consequence of the non-covalent nature of their core interactions, but also afford unique structural characteristics. Although much is now known about the manner in which such structures adopt their morphologies and size distributions in response to external stimuli, the kinetic and thermodynamic driving forces that lead to their transformation from soluble monomeric species into ordered supramolecular entities have remained elusive. Here we focus on Boc-diphenylalanine, an archetypical example of a peptide with a high propensity towards supramolecular self-organization, and describe the pathway through which it forms a range of nano-assemblies with different structural characteristics. Our results reveal that the nucleation process is multi-step in nature and proceeds by Ostwald's step rule through which coalescence of soluble monomers leads to the formation of nanospheres, which then undergo ripening and structural conversions to form the final supramolecular assemblies. We characterize the structures and thermodynamics of the different phases involved in this process and reveal the intricate nature of the transitions that can occur between discrete structural states of this class of supramolecular polymers.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard