MilliporeSigma
  • A gas chromatography-mass spectrometry method for the determination of pogostone in canine plasma and its application to a pharmacokinetic study.
A gas chromatography-mass spectrometry method for the determination of pogostone in canine plasma and its application to a pharmacokinetic study.
Journal of chromatographic science (2014-04-10)
Ruoqi Zhang, Cheng Peng, Yunxia Li, Liang Xiong, Xiaohong Gong
ABSTRACT
In this study, a simple and selective gas chromatography-mass spectrometry method was developed and validated for the determination of pogostone in canine plasma. Liquid-liquid extraction was used to separate pogostone from canine plasma, and the mean extraction recovery rates of pogostone and the internal standard (isoalantolactone) were 80.61 and 75.89%, respectively. Plastic centrifuge tubes were inadequate for the plasma sample treatment procedure because of the adsorption effect of pogostone on the inner surface. The chromatographic separation was performed on a capillary column of Agilent HP-5ms, and the spectrometer was operated in an electron-impact ionization with an electron multiplier voltage mode. The standard curve was linear over the concentration range of 1.02-406 ng/mL (r > 0.99). The intra- and interday accuracies for pogostone at four concentrations were 97.77-99.92% and 98.51-100.22%, respectively. The relative standard deviations were <15%. The method was successfully applied to a pharmacokinetic study after the oral administration of pogostone to beagle dogs.
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%