MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Hyperglycaemia enhances nitric oxide production in diabetes: a study from South Indian patients.
Hyperglycaemia enhances nitric oxide production in diabetes: a study from South Indian patients.
PloS one (2015-04-22)
Ramu Adela, Susheel Kumar Nethi, Pankaj K Bagul, Ayan K Barui, Saidulu Mattapally, Madhusudan Kuncha, Chitta R Patra, P Naveen Chander Reddy, Sanjay K Banerjee
ABSTRACT
We have previously reported that increased glucose levels were associated with higher serum nitric oxide (NO) levels in fructose-fed insulin resistant rats. However, the relationship between hyperglycemia and serum NO level was not clear. Therefore, the present study was designed to find the association between hyperglycemia and serum NO levels in Type 2 diabetic (T2DM) patients and T2DM with cardiovascular complication. Endothelial cells (HUVEC) were treated with of D-glucose (10-100mM), and NO levels and NOS gene expression was measured. Hyperglycaemia was induced in Sprague-Dawley rats, and serum NO levels were measured after 8 weeks. For clinical evaluation, five groups of patients were recruited: Control (CT, n=48), Type 2 diabetes (T2DM, n=26), T2DM with hypertension (DMHT, n=46), Coronary artery diseases (CAD, n=29) and T2DM with coronary artery diseases (DMCD, n=38). NO (nitrite + nitrate) levels were measured from human serum. We found a significant (p<0.05) and dose-dependent increase in NO levels in HUVEC cells after 4 hours of high glucose exposure. eNOS and iNOS gene expression was increased in HUVEC cells after different concentrations and time periods of glucose treatment. We also observed significant (149.1 ± 25 μM, p<0.01) increase in serum NO levels in hyperglycaemic rats compared to control (76.6 ± 13.2 μM). Serum NO level was significantly higher in T2DM (111.8 μM (81.7-122.4), p<0.001) and DMCD patients ((129.4 μM (121.2-143.5), p <0.001) but not in CAD patients (76.4 μM (70.5-87)), as compared to control (68.2 μM (56.4-82.3)). We found significantly lower NO levels (83.5 μM (60.5-122.9)) in subjects suffering from diabetes since more than 5 years, compared to subjects (115.3 μM (75.2-127.1), p<0.001) with less than 5 years. In conclusion, high NO levels were observed in South Indian diabetic patients. Higher glucose levels in serum might be responsible for activation of endothelial cells to enhance NO levels.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard