MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Mst1 inhibition rescues β1-adrenergic cardiomyopathy by reducing myocyte necrosis and non-myocyte apoptosis rather than myocyte apoptosis.
Mst1 inhibition rescues β1-adrenergic cardiomyopathy by reducing myocyte necrosis and non-myocyte apoptosis rather than myocyte apoptosis.
Basic research in cardiology (2015-01-21)
Grace J Lee, Lin Yan, Dorothy E Vatner, Stephen F Vatner
ABSTRACT
It is generally held that inhibition of mammalian sterile 20-like kinase 1 (Mst1) protects the heart through reducing myocyte apoptosis. We determined whether inhibition with a dominant-negative Mst1 (DN-Mst1) would protect against the cardiomyopathy induced by chronic β1-adrenergic receptor (β1-AR) stimulation by preventing myocyte apoptosis. DN-Mst1 mice were mated with β1-AR transgenic (Tg) mice and followed for 20 months. β1-AR Tg mice developed cardiomyopathy as they aged, as reflected by premature mortality and depressed cardiac function, which were rescued in β1-AR × DN-Mst1 bigenic mice. Surprisingly, myocyte apoptosis did not significantly decrease with Mst1 inhibition. Instead, Mst1 inhibition predominantly reduced non-myocyte apoptosis, e.g., fibroblasts, macrophages, neutrophils and endothelial cells. Fibrosis in the hearts with cardiomyopathy increased fivefold and this increase was nearly abolished in the bigenic mice with Mst1 inhibition. Regression analysis showed no correlation between myocyte apoptosis and cardiac function or myocyte number, whereas the latter two correlated significantly, p < 0.05, with fibrosis, which generally results from necrosis. To examine the role of myocyte necrosis, chronic β-AR stimulation with isoproterenol was induced for 24 h and myocyte necrosis was assessed by 1% Evans blue dye. Compared to WT, DN-Mst1 mice showed significant inhibition, p < 0.05, of myocyte necrosis. We confirmed this result in Mst1-knockout mice, which also showed significant protection, p < 0.05, against myocyte necrosis compared to WT. These data indicate that Mst1 inhibition rescued cardiac fibrosis and myocardial dysfunction in β1-AR cardiomyopathy. However, this did not occur through Mst1 inhibition of myocyte apoptosis but rather by inhibition of cardiomyocyte necrosis and non-myocyte apoptosis, features of Mst1 not considered previously.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)