MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Doxycycline as an inhibitor of the epithelial-to-mesenchymal transition and vasculogenic mimicry in hepatocellular carcinoma.
Doxycycline as an inhibitor of the epithelial-to-mesenchymal transition and vasculogenic mimicry in hepatocellular carcinoma.
Molecular cancer therapeutics (2014-10-04)
Jie Meng, Baocun Sun, Xiulan Zhao, Danfang Zhang, Xueming Zhao, Qiang Gu, Xueyi Dong, Nan Zhao, Peimei Liu, Yanrong Liu
ABSTRACT
This study was conducted to examine the effects of doxycycline on the survival time and proliferation of hepatocellular carcinoma (HCC) in vivo and on the biologic functions of HCC in vitro. This study was also designed to evaluate the effects of doxycycline on epithelial-to-mesenchymal transition (EMT)- and vasculogenic mimicry (VM)-related protein expression and on matrix metalloproteinase (MMP) and DNA methyltransferase (DNMT) activity in vitro. Human MHCC97H cells were injected into BALB/c mice, which were divided into treatment and control groups. Doxycycline treatment prolonged the mouse survival time and partly suppressed the growth of engrafted HCC tumor cells, with an inhibition rate of 43.39%. Higher amounts of VM and endothelium-dependent vessels were found in the control group than the treatment group. IHC indicated that epithelial (E)-cadherin expression was increased in the doxycycline-treated mice compared with the control group. In in vitro experiments, doxycycline promoted HCC cell adhesion but inhibited HCC cell viability, proliferation, migration, and invasion. Western blot analysis, semiquantitative RT-PCR, qRT-PCR, and immunofluorescence demonstrated that doxycycline inhibited the degradation of the epithelial marker E-cadherin and downregulated the expression levels of EMT promoters, the mesenchymal marker vimentin, and the VM-associated marker vascular endothelial (VE)-cadherin. Furthermore, the activities of MMPs and DNMTs were examined in different groups via gelatin zymography and a DNMT activity assay kit. A methylation-specific PCR was performed to assess the promoter methylation of CDH1 (the gene encoding E-cadherin). Doxycycline prolonged the mouse survival time by inhibiting EMT progression and VM formation.
Product Description
Supelco
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydroquinone, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethidium bromide, ≥95.0%
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Sodium hydrogen sulfate
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL