MilliporeSigma
  • A high-throughput robotic sample preparation system and HPLC-MS/MS for measuring urinary anatabine, anabasine, nicotine and major nicotine metabolites.
A high-throughput robotic sample preparation system and HPLC-MS/MS for measuring urinary anatabine, anabasine, nicotine and major nicotine metabolites.
Clinica chimica acta; international journal of clinical chemistry (2014-06-27)
Binnian Wei, June Feng, Imran J Rehmani, Sharyn Miller, James E McGuffey, Benjamin C Blount, Lanqing Wang
ABSTRACT
Most sample preparation methods characteristically involve intensive and repetitive labor, which is inefficient when preparing large numbers of samples from population-scale studies. This study presents a robotic system designed to meet the sampling requirements for large population-scale studies. Using this robotic system, we developed and validated a method to simultaneously measure urinary anatabine, anabasine, nicotine and seven major nicotine metabolites: 4-Hydroxy-4-(3-pyridyl)butanoic acid, cotinine-N-oxide, nicotine-N-oxide, trans-3'-hydroxycotinine, norcotinine, cotinine and nornicotine. We analyzed robotically prepared samples using high-performance liquid chromatography (HPLC) coupled with triple quadrupole mass spectrometry in positive electrospray ionization mode using scheduled multiple reaction monitoring (sMRM) with a total runtime of 8.5 min. The optimized procedure was able to deliver linear analyte responses over a broad range of concentrations. Responses of urine-based calibrators delivered coefficients of determination (R(2)) of >0.995. Sample preparation recovery was generally higher than 80%. The robotic system was able to prepare four 96-well plate (384 urine samples) per day, and the overall method afforded an accuracy range of 92-115%, and an imprecision of <15.0% on average. The validation results demonstrate that the method is accurate, precise, sensitive, robust, and most significantly labor-saving for sample preparation, making it efficient and practical for routine measurements in large population-scale studies such as the National Health and Nutrition Examination Survey (NHANES) and the Population Assessment of Tobacco and Health (PATH) study.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O