MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Atorvastatin-mediated protection of the retina in a model of diabetes with hyperlipidemia.
Atorvastatin-mediated protection of the retina in a model of diabetes with hyperlipidemia.
Canadian journal of physiology and pharmacology (2014-11-19)
Rosa Fernandes, Carla F Bento, Paulo Matafome, Cristina M Sena, Raquel M Seiça, Paulo Pereira
ABSTRACT
Insulin resistance, a key feature of obesity, metabolic syndrome, and type 2 diabetes mellitus (T2DM), results in a variety of metabolic and vascular abnormalities. Metabolic disturbances associated with diabetes could contribute to disrupting the structural and (or) functional integrity of the retina. The effects of atorvastatin on retinal cells in hyperlipidemic T2DM rats have not yet been investigated. We used Goto-Kakizaki (GK) rats fed with an atherogenic diet (AD) for 4 months to investigate whether atorvastatin (administered for 1 month) would slow-down or reverse the progression of lesions in the diabetic retina. Fluorogenic substrates were used to measure the proteasome activities in retinal cells. The production of reactive oxygen species was determined by immunofluorescence in frozen retina sections, using dihydroethydium. Nitrotyrosine levels were assessed using immunohistochemistry. Protein levels of ubiquitin conjugates, free ubiquitin, and ubiquitin activating enzyme E1 were determined with Western blotting. Atorvastatin significantly reduced the levels of oxidative stress that were induced by the AD and restored the proteasome activities in the diabetic GK rats. Atorvastatin therapy significantly improved local oxidative stress levels in GK rats fed with AD. Atorvastatin can, at least in part, restore the ubiquitin proteasome system, and may represent a pharmacological approach to prevent some of the complications associated with diabetic retinopathy.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)
Sigma-Aldrich
6-Maleimidohexanoic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
SAFC
Iodoacetamide
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, BioUltra