MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Alginate encapsulant incorporating CXCL12 supports long-term allo- and xenoislet transplantation without systemic immune suppression.
Alginate encapsulant incorporating CXCL12 supports long-term allo- and xenoislet transplantation without systemic immune suppression.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2015-02-20)
T Chen, J Yuan, S Duncanson, M L Hibert, B C Kodish, G Mylavaganam, M Maker, H Li, M Sremac, M Santosuosso, B Forbes, S Kashiwagi, J Cao, J Lei, M Thomas, C Hartono, D Sachs, J Markmann, A Sambanis, M C Poznansky
ABSTRACT
Islet transplantation represents a potentially curative approach for individuals with Type I Diabetes. The requirement for systemic immune suppression to control immune-mediated rejection of transplanted islets and the limited human islet supply represent significant roadblocks to progress for this approach. Islet microencapsulation in alginate offers limited protection in the absence of systemic immunosuppression, but does not support long-term islet survival. The chemokine, CXCL12, can repel effector T cells while recruiting immune-suppressive regulatory T cells (Tregs) to an anatomic site while providing a prosurvival signal for beta-cells. We proposed that coating or encapsulating donor islets with CXCL12 would induce local immune-isolation and protect and support the function of an allo- or xenograft without systemic immune suppression. This study investigated the effect of alginate microcapsules incorporating CXCL12 on islet function. Islet transplantation was performed in murine models of insulin-dependent diabetes. Coating of islets with CXCL12 or microencapsulation of islets with alginate incorporating the chemokine, resulted in long-term allo- and xenoislet survival and function, as well as a selective increase in intragraft Tregs. These data support the use of CXCL12 as a coating or a component of an alginate encapsulant to induce sustained local immune-isolation for allo- or xenoislet transplantation without systemic immunosuppression.
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
SAFC
L-Glutamine
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM