MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Multitarget effects of quercetin in leukemia.
Cancer prevention research (Philadelphia, Pa.) (2014-10-09)
Victor Maso, Andrana Karla Calgarotto, Gilberto Carlos Franchi, Alexandre Eduardo Nowill, Paulo Latuf Filho, José Vassallo, Sara Teresinha Olalla Saad
ABSTRACT
This study proposes to investigate quercetin antitumor efficacy in vitro and in vivo, using the P39 cell line as a model. The experimental design comprised leukemic cells or xenografts of P39 cells, treated in vitro or in vivo, respectively, with quercetin; apoptosis, cell-cycle and autophagy activation were then evaluated. Quercetin caused pronounced apoptosis in P39 leukemia cells, followed by Bcl-2, Bcl-xL, Mcl-1 downregulation, Bax upregulation, and mitochondrial translocation, triggering cytochrome c release and caspases activation. Quercetin also induced the expression of FasL protein. Furthermore, our results demonstrated an antioxidant activity of quercetin. Quercetin treatment resulted in an increased cell arrest in G1 phase of the cell cycle, with pronounced decrease in CDK2, CDK6, cyclin D, cyclin E, and cyclin A proteins, decreased Rb phosphorylation and increased p21 and p27 expression. Quercetin induced autophagosome formation in the P39 cell line. Autophagy inhibition induced by quercetin with chloroquine triggered apoptosis but did not alter quercetin modulation in the G1 phase. P39 cell treatment with a combination of quercetin and selective inhibitors of ERK1/2 and/or JNK (PD184352 or SP600125, respectively), significantly decreased cells in G1 phase, this treatment, however, did not change the apoptotic cell number. Furthermore, in vivo administration of quercetin significantly reduced tumor volume in P39 xenografts and confirmed in vitro results regarding apoptosis, autophagy, and cell-cycle arrest. The antitumor activity of quercetin both in vitro and in vivo revealed in this study, point to quercetin as an attractive antitumor agent for hematologic malignancies.
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
PD184352, ≥98% (HPLC)
Sigma-Aldrich
Mitochondria/Cytosol Fractionation Kit, This Mitochondria/Cytosol Fractionation Kit provides reagents for quick & efficient isolation of intact mitochondria from cultured cells.
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
tert-Butyl hydroperoxide solution, CP
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard