MilliporeSigma
  • Peripapillary rat sclera investigated in vivo with polarization-sensitive optical coherence tomography.
Peripapillary rat sclera investigated in vivo with polarization-sensitive optical coherence tomography.
Investigative ophthalmology & visual science (2014-10-30)
Bernhard Baumann, Sabine Rauscher, Martin Glösmann, Erich Götzinger, Michael Pircher, Stanislava Fialová, Marion Gröger, Christoph K Hitzenberger
ABSTRACT
To demonstrate polarization-sensitive (PS) optical coherence tomography (OCT) for noninvasive, volumetric, and quantitative imaging of the birefringent properties of the peripapillary rat sclera; to compare the findings from PS-OCT images to state-of-the-art histomorphometric analysis of the same tissues. A high-speed PS-OCT prototype operating at 840 nm was modified for imaging the rat eye. Densely sampled PS-OCT raster scans covering an area of ~1.5 × 1.5 mm centered at the papilla were acquired in the eyes of anesthetized male Sprague-Dawley rats. Cross-sectional PS-OCT images were computed, and fundus maps displaying the birefringent properties of the sclera were analyzed. Postmortem histomorphologic analysis was performed. Polarization-sensitive OCT enables visualization of the polarization properties of ocular tissues in vivo. The birefringent characteristics of the rat sclera were quantitatively assessed. Scleral birefringence formed a donut-shaped pattern around the papilla with significantly increased values of 0.703 ± 0.089°/μm (i.e., 1.64 × 10(-3) ± 0.2 × 10(-3); mean ± standard deviation) and 0.721 ± 0.084°/μm (i.e., 1.68 × 10(-3) ± 0.2 × 10(-3)) at an eccentricity of 0.4 mm for the left and right eyes, respectively. Birefringent axis orientation maps revealed a ring-shaped distribution around the optic nerve. Postmortem PS-OCT micrographs provided access to retinal and scleral microstructure and were compared to standard histomorphologic analysis. Polarization-sensitive OCT enables quantitative imaging of tissue polarization properties in addition to conventional OCT imaging based on reflectivity. In the rat sclera, in vivo PS-OCT provides access to volumetric mapping of birefringence. Scleral birefringence is associated with microstructural tissue organization. Therefore, PS-OCT should prove a valuable tool for the in vivo investigation of peripapillary sclera in glaucoma.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications