MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Coordination of the filament stabilizing versus destabilizing activities of cofilin through its secondary binding site on actin.
Coordination of the filament stabilizing versus destabilizing activities of cofilin through its secondary binding site on actin.
Cytoskeleton (Hoboken, N.J.) (2014-06-20)
Dimitra Aggeli, Erik Kish-Trier, Meng Chi Lin, Brian Haarer, Gino Cingolani, John A Cooper, Stephan Wilkens, David C Amberg
ABSTRACT
Cofilin is a ubiquitous modulator of actin cytoskeleton dynamics that can both stabilize and destabilize actin filaments depending on its concentration and/or the presence of regulatory co-factors. Three charge-reversal mutants of yeast cofilin, located in cofilin's filament-specific secondary binding site, were characterized in order to understand why disruption of this site leads to enhanced filament disassembly. Crystal structures of the mutants showed that the mutations specifically affect the secondary actin-binding interface, leaving the primary binding site unaltered. The mutant cofilins show enhanced activity compared to wild-type cofilin in severing and disassembling actin filaments. Electron microscopy and image analysis revealed long actin filaments in the presence of wild-type cofilin, while the mutants induced many short filaments, consistent with enhanced severing. Real-time fluorescence microscopy of labeled actin filaments confirmed that the mutants, unlike wild-type cofilin, were functioning as constitutively active severing proteins. In cells, the mutant cofilins delayed endocytosis, which depends on rapid actin turnover. We conclude that mutating cofilin's secondary actin-binding site increases cofilin's ability to sever and de-polymerize actin filaments. We hypothesize that activators of cofilin severing, like Aip1p, may act by disrupting the interface between cofilin's secondary actin-binding site and the actin filament.
Product Description
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Supelco
Pyrene, analytical standard
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%