MilliporeSigma
  • The major cow milk allergen Bos d 5 manipulates T-helper cells depending on its load with siderophore-bound iron.
The major cow milk allergen Bos d 5 manipulates T-helper cells depending on its load with siderophore-bound iron.
PloS one (2014-08-15)
Franziska Roth-Walter, Luis F Pacios, Cristina Gomez-Casado, Gerlinde Hofstetter, Georg A Roth, Josef Singer, Araceli Diaz-Perales, Erika Jensen-Jarolim
ABSTRACT
The mechanisms of allergic sensitization to milk are still elusive. The major allergen Bos d 5 belongs to the lipocalin-family and thus is able to transport numerous ligands. In this study we investigated its ability to bind to iron-siderophore complexes and tested the immune-modulatory properties of Bos d 5 in either forms. Structural and in silico docking analysis of Bos d 5 revealed that Bos d 5 is able to bind to iron via catechol-based flavonoids (quercetin, myricetin, luteolin) that act as siderophores as confirmed by spectral-analysis and iron staining. Calculated dissociation constants of docking analyses were below 1 µM by virtual addition of iron. When incubated with human peripheral blood mononuclear cells (PBMCs), only the apo-form of Bos d 5 led to an increase of CD4+positive cells and significantly elevated IL13 and IFNγ-levels. In contrast, holo-Bos d 5 decreased numbers of CD4 expressing cells and induced apoptosis. Taken together, our data give evidence that Bos d 5 is capable of binding iron via siderophores. Moreover, our data support for the first time the notion that the form of application (apo- or holo-form) is decisive for the subsequent immune response. The apo-form promotes Th2 cells and inflammation, whereas the holo-form appears to be immunosuppressive.
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Human IFNA1 / Interferon Alpha-1/13 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Human IFN γ ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IFN γ ELISA Kit, for cell and tissue lysates
Natural Bos d 5 (β-Lactoglobulin), Cow′s milk, pkg of 250 μg, storage temp.:-20°C
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)