MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • One-pot synthesis of C₁₈-functionalized core-shell magnetic mesoporous silica composite as efficient sorbent for organic dye.
One-pot synthesis of C₁₈-functionalized core-shell magnetic mesoporous silica composite as efficient sorbent for organic dye.
Journal of colloid and interface science (2015-03-04)
Xiaole Zhang, Tao Zeng, Saihua Wang, Hongyun Niu, Xiaoke Wang, Yaqi Cai
ABSTRACT
In this work, a facile one-pot strategy was proposed for the synthesis of C18-functionalized core-shell magnetic mesoporous silica composite (Fe3O4/mSiO2-C18). The Fe3O4/mSiO2-C18 composite, with an average size of 80 nm and a functionalized mesoporous silica shell of about 30 nm in thickness, has excellent adsorption ability toward methylene blue dye (MB) due to the large surface area (303 m(2) g(-1)) and the abundant hydrophobic C18 groups. The adsorption equilibrium was achieved within 20 min and the adsorption behavior of MB on Fe3O4/mSiO2-C18 composite fitted the pseudo-second-order kinetic model well (k2=1.29×10(-2) g mg(-1) min(-1), q(e)=144.72 mg g(-1), h(o)=270.27 mg g(-1) min(-1) under 25 °C and an initial MB concentration of 10 mg L(-1)). Langmuir and Freundlich isothermal adsorption models can both be used to describe the adsorption process and the maximum Langmuir adsorption capacity of MB on Fe3O4/mSiO2-C18 at 25 °C and pH 7.5 is 363.64 mg g(-1). Thermodynamic parameters show that the adsorption reaction is exothermic and spontaneous (ΔH(0)=-63.49 kJ mol(-1), ΔG(0)=-7.80 kJ mol(-1)). Ionic strength and pH affected the adsorption slightly. In addition, the MB adsorbed sorbent can be readily separated from water solution by an external magnet because of the high magnetic saturation value (22.62 emu g(-1)). After being regenerated by treatment with acidic methanol, the sorbent could be reused for at least 5 cycles with a little decrease in adsorption capacity.
Sigma-Aldrich
Tetraethyl orthosilicate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
n-Octadecyltriethoxysilane, AldrichCPR
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methylene Blue solution
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000