MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Monitoring Glycan-Protein Interactions by NMR Spectroscopic Analysis: A Simple Chemical Tag That Mimics Natural CH-π Interactions.
Monitoring Glycan-Protein Interactions by NMR Spectroscopic Analysis: A Simple Chemical Tag That Mimics Natural CH-π Interactions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-17)
Luis P Calle, Begoña Echeverria, Antonio Franconetti, Sonia Serna, M Carmen Fernández-Alonso, Tammo Diercks, F Javier Cañada, Ana Ardá, Niels-Christian Reichardt, Jesús Jiménez-Barbero
ABSTRACT
Detection of molecular recognition processes requires robust, specific, and easily implementable sensing methods, especially for screening applications. Here, we propose the difluoroacetamide moiety (an acetamide bioisoster) as a novel tag for detecting by NMR analysis those glycan-protein interactions that involve N-acetylated sugars. Although difluoroacetamide has been used previously as a substituent in medicinal chemistry, here we employ it as a specific sensor to monitor interactions between GlcNAc-containing glycans and a model lectin (wheat germ agglutinin). In contrast to the widely employed trifluoroacetamide group, the difluoroacetamide tag contains geminal (1) H and (19) F atoms that allow both (1) H and (19) F NMR methods for easy and robust detection of molecular recognition processes involving GlcNAc- (or GalNAc-) moieties over a range of binding affinities. The CHF2 CONH- moiety behaves in a manner that is very similar to that of the natural acetamide fragment in the involved aromatic-sugar interactions, providing analogous binding energy and conformations, whereas the perfluorinated CF3 CONH- analogue differs more significantly.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG