MilliporeSigma
  • Individual and combined cytotoxic effects of aflatoxin B1, zearalenone, deoxynivalenol and fumonisin B1 on BRL 3A rat liver cells.
Individual and combined cytotoxic effects of aflatoxin B1, zearalenone, deoxynivalenol and fumonisin B1 on BRL 3A rat liver cells.
Toxicon : official journal of the International Society on Toxinology (2015-01-01)
Lv-Hui Sun, Ming-Yan Lei, Ni-Ya Zhang, Xin Gao, Chong Li, Christopher Steven Krumm, De-Sheng Qi
ABSTRACT
This study was performed to determine the individual and combined cytotoxic effects of Aflatoxin B1 (AFB1), zearalenone (ZEA), deoxynivalenol (DON) and fumonisin B1 (FB1) on BRL 3A rat liver cells. After the mycotoxins treated the BRL 3A cells for 12, 24 and 48 h, cell viability was determined using the MTT assay. The cytotoxicity of individual mycotoxins on BRL 3A cell viability in decreasing order were DON > AFB1 > ZEA > FB1. The central composite design (CCD) was used to assess the toxicity of binary and ternary mixtures of these mycotoxins. The mixtures of AFB1+ZEA and AFB1+DON showed the synergetic toxic effects on BRL 3A cells. These toxins decreased the viability of cells by inducing intracellular reactive oxygen species (ROS) production and promoting apoptosis in the BRL 3A cells. This effect was mediated by an upregulation of the stress and apoptotic genes Hsp70, p53, Bax, Caspase-3 and Caspase-8, along with a downregulation of the antiapoptotic gene Bcl-2. In conclusion, our results suggested that the coexistence of AFB1 and ZEA or DON in agricultural products could be more hepatotoxic than individually, suggests that the toxicological interactions of these toxins need to be better understood to assess health risks.
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder