MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A core-shell-structured molecularly imprinted polymer on upconverting nanoparticles for selective and sensitive fluorescence sensing of sulfamethazine.
A core-shell-structured molecularly imprinted polymer on upconverting nanoparticles for selective and sensitive fluorescence sensing of sulfamethazine.
The Analyst (2015-06-16)
Jinghan Tian, Jialei Bai, Yuan Peng, Zhiwei Qie, Yufeng Zhao, Baoan Ning, Dan Xiao, Zhixian Gao
ABSTRACT
A core-shell structured molecularly imprinted polymer on upconverting nanoparticles (UCNPs@MIP) was synthesized for the fluorescence (FL) sensing of sulfamethazine (SMZ). Hexagonal UCNPs were synthesized by the solvothermal method, then coated with a thin silica shell and modified with vinyl groups. Finally, surface polymerization was initiated among the vinyl groups, the functional monomers and cross-linking agents by the initiator. The MIP synthesized by this procedure was anchored on the surface of UCNPs, possessed better site accessibility and lower transfer resistance for the target molecule compared to bulk imprinted materials. The obtained UCNPs@MIP showed good binding capacity, fast response, high selectivity and specificity to the SMZ. The FL intensity of the UCNPs@MIP decreased sensitively with the increasing concentration of SMZ in the range of 50-700 ng mL(-1), the detection limit was 34 ng mL(-1) (S/N = 3). The UCNPs@MIP was successfully applied to the detection of SMZ in chicken samples. Thanks to the unique near-infrared (NIR) excitation nature of UCNPs, the chicken meat only needed some simple extraction procedures before FL detection, no complex purifications were required. The average recoveries ranged from 96.01% to 98.90%, with relative standard deviations (RSDs) below 4.5%. This work offers a novel sensing system that combined the advantages of upconverting nanotechnology and molecularly imprinted technology.
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Tetraethyl orthosilicate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis