MilliporeSigma
  • Radiotherapy-induced hypopituitarism in nasopharyngeal carcinoma: the tip of an iceberg.
Radiotherapy-induced hypopituitarism in nasopharyngeal carcinoma: the tip of an iceberg.
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association (2015-05-27)
S H Ipekci, M Cakir, A Kiyici, O Koc, M Artac
ABSTRACT
Radiation-induced hypopituitarism is an important late complication of cranial radiotherapy in children and adults. The purpose of this cross-sectional study was to evaluate the effects of radiotherapy on pituitary function in adult nasopharyngeal carcinoma patients. Pituitary function was evaluated in 30 patients after cranial radiotherapy for nasopharyngeal carcinoma. Somatotroph and corticotroph axes were assessed by insulin tolerance test while gonadotroph and thyroid axes were evaluated by basal pituitary and end organ hormone levels at 10-133 months after radiotherapy. At least one hormonal disorder was observed in 28 (93%) patients after radiotherapy. 26 (87%) patients had one or more anterior pituitary hormone deficiencies. The rates of pituitary hormone deficiencies were 77% for growth hormone, followed by adrenocorticotropic hormone (73%), thyroid-stimulating hormone (27%) and gonadotropins (7%). Hyperprolactinemia was present in 13 (43%) patients. Radiation-induced hypopituitarism is more common than expected in patients with nasopharyngeal carcinoma.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®