MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Alteplase Reduces Downstream Microvascular Thrombosis and Improves the Benefit of Large Artery Recanalization in Stroke.
Alteplase Reduces Downstream Microvascular Thrombosis and Improves the Benefit of Large Artery Recanalization in Stroke.
Stroke (2015-10-08)
Jean-Philippe Desilles, Stephane Loyau, Varouna Syvannarath, Jaime Gonzalez-Valcarcel, Marie Cantier, Liliane Louedec, Bertrand Lapergue, Pierre Amarenco, Nadine Ajzenberg, Martine Jandrot-Perrus, Jean-Baptiste Michel, Benoit Ho-Tin-Noe, Mikael Mazighi
ABSTRACT
Downstream microvascular thrombosis (DMT) is known to be a contributing factor to incomplete reperfusion in acute ischemic stroke. The aim of this study was to determine the timing of DMT with intravital imaging and to test the hypothesis that intravenous alteplase infusion could reduce DMT in a transient middle cerebral artery occlusion (MCAO) rat stroke model. Rats were subjected to 60-minute transient MCAO. Alteplase (10 mg/kg) was administered 30 minutes after the beginning of MCAO. Real-time intravital fluorescence microscopy through a dura-sparing craniotomy was used to visualize circulating blood cells and fibrinogen. Cerebral microvessel patency was quantitatively evaluated by fluorescein isothiocyanate-dextran perfusion. Immediately after MCAO, platelet and leukocyte accumulation were observed mostly in the venous compartment. Within 30 minutes after MCAO, microthrombi and parietal fibrin deposits were detected in postcapillary microvessels. Alteplase treatment significantly (P=0.006) reduced infarct volume and increased the percentage of perfused vessels during MCAO (P=0.02) compared with saline. Plasma levels of fibrinogen from alteplase-treated rats showed a rapid and profound hypofibrinogenemia. In vitro platelet aggregation demonstrated that alteplase reduced platelet aggregation (P=0.0001) and facilitated platelet disaggregation (P=0.001). These effects were reversible in the presence of exogenous fibrinogen. Our data demonstrate that DMT is an early phenomenon initiated before recanalization. We further show that alteplase-dependent maintenance of downstream perfusion during MCAO improves acute ischemic stroke outcome through a fibrinogen-dependent platelet aggregation reduction. Our results indicate that early targeting of DMT represents a therapeutic strategy to improve the benefit of large artery recanalization in acute ischemic stroke.
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, JIS special grade, 98.0-102.0%
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)