MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Dissolution of biological samples in deep eutectic solvents: an approach for extraction of polycyclic aromatic hydrocarbons followed by liquid chromatography-fluorescence detection.
Dissolution of biological samples in deep eutectic solvents: an approach for extraction of polycyclic aromatic hydrocarbons followed by liquid chromatography-fluorescence detection.
Journal of chromatography. A (2015-04-11)
Zahra Helalat-Nezhad, Kamal Ghanemi, Mehdi Fallah-Mehrjardi
ABSTRACT
A novel sample preparation method based on the complete dissolution of marine biological samples in choline chloride-oxalic acid (ChCl-Ox) deep eutectic solvent was developed for fast and efficient extraction of eight polycyclic aromatic hydrocarbons (PAHs) using minimum volumes of cyclohexane. The extracted PAHs were purified and then measured by high-performance liquid chromatography-fluorescence detection (HPLC-FL). The effect of key parameters on extraction recoveries and precision was investigated. At optimized conditions, the studied samples were dissolved under atmospheric pressure in ChCl-Ox (1:2) at 55°C for 30min, which is considerably lower than the temperature used in the classical and current methods. After dissolution, it took approximately 20min to quantitatively extract the PAHs from ChCl-Ox using 5mL cyclohexane. Depending on the analyte, the developed method was linear over the calibration range 1.0-250, 2.0-250, and 5.0-250ngg(-1), with r(2)>0.996. The detection limits of the method were between 0.50 and 3.08ngg(-1). The intra-day and inter-day precisions (based on the relative standard deviation, n=5) of the spiked PAHs at a concentration level of 50ngg(-1) were better than 12.6% and 13.3%, respectively. Individual PAH recoveries from spiked marine fish and macroalgae samples were in the range of 71.6% to 109.6%. For comparison, the spiked samples were also subjected to the Soxhlet extraction method. The simplicity of the procedure, high extraction efficiency, short analysis time, and use of safe and inexpensive components suggest the proposed method has a high potential for utilization in routine trace PAH analysis in biological samples.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Choline chloride, ≥99%
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Toluene, JIS 1000, suitable for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, JIS 300, suitable for residue analysis, ≥99.8%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Supelco
Dichloromethane, ≥99.9%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, suitable for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, suitable for residue analysis
Sigma-Aldrich
Hexane, suitable for HPLC