MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Co-extraction and simultaneous determination of multi-class hydrophobic organic contaminants in marine sediments and biota using GC-EI-MS/MS and LC-ESI-MS/MS.
Co-extraction and simultaneous determination of multi-class hydrophobic organic contaminants in marine sediments and biota using GC-EI-MS/MS and LC-ESI-MS/MS.
Talanta (2015-06-17)
Hui Zhang, Stéphane Bayen, Barry C Kelly
ABSTRACT
A multi-residue analytical method was developed involving co-extraction and simultaneous determination of 89 hydrophobic organic contaminants (HOCs) in marine sediments and biota using gas chromatography-electron ionization-triple quadrupole mass spectrometry (GC-EI-MS/MS) and liquid chromatography-electrospray ionization-triple quadrupole mass spectrometry (LC-ESI-MS/MS). Target analytes include polychlorinated biphenyls (PCBs), pesticides, chlorobenzenes, brominated and chlorinated flame retardants, nitro-aromatic and polycyclic musks, triclosan and methyl triclosan. Spike-recovery experiments showed relative recoveries of analytes were generally between 70% and 130%. Analyses of a sediment standard reference material (SRM 1944) demonstrated recoveries between 80% and 120% for certified concentrations of individual PCBs and pesticides. Method detection limits of individual compounds ranged from 0.1 to 57.1 pg/g dw for sediments and 0.1 to 22.8 pg/g ww for biota. A field survey of sediments and biota from Singapore's marine environment demonstrated the occurrence of polycyclic and nitro-aromatic musks (galaxolide, tonalide, musk ketone, etc.), halogenated flame retardants (syn- and anti-dechlorane plus (DP), α, β and γ-hexabromocyclododecane (HBCD), tetrabromobisphenol A (TBBPA), etc.), as well as triclosan and methyl triclosan. Galaxolide exhibited relatively high concentrations, with highest levels in polychaete worms (161.7±72.5 ng/g ww) and clams (546.8±220.3 ng/g ww) from mangroves. Triclosan and methyl triclosan levels were highly correlated in sediments (r(2)=0.9752), while syn- and anti-DP were strongly correlated in biota (r(2)=0.9279). anti-DP/syn-DP stereoisomer ratios were typically >1 and ranged between 0.94 and 29.2 in sediments and biota samples. γ-HBCD exhibited the highest concentrations among HBCD isomers in sediments. Conversely, α-HBCD was the dominant HBCD isomer in biota.
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
1,3,5-Tribromobenzene, 98%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
1,2,4,5-Tetrabromobenzene, 97%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Dichloromethane, ≥99.9%