MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Aprotinin revisited: formulation, characterization, biodistribution and therapeutic potential of new aprotinin microemulsion in acute pancreatitis.
Aprotinin revisited: formulation, characterization, biodistribution and therapeutic potential of new aprotinin microemulsion in acute pancreatitis.
Journal of drug targeting (2015-03-05)
H Yeşim Karasulu, Nevin Oruç, Neslihan Üstündağ-Okur, Derya İlem Özdemir, Zeynep Ay Şenyiğit, Funda Barbet Yılmaz, Makbule Aşıkoğlu, Hayal Özkılıç, Eren Akçiçek, Tamer Güneri, Ömer Özütemiz
ABSTRACT
The aim of this study was to develop aprotinin-loaded microemulsion (MA) for intravenous administration and evaluate the biodistribution and therapeutic potential of developed formulation in acute pancreatitis models in rats. Phase diagrams were constructed to identify microemulsion region and the optimal microemulsion was evaluated for physicochemical properties and treatment effect in rats, and comparisons made with the solution of aprotinin (SA). To evaluate the biodistribution of the drug by gamma scintigraphy aprotinin was radiolabeled with (99m)Tc radionuclide. Mild and severe acute pancreatitis was induced in rats by subcutaneous injections of cerulein and introductal infusion of 3% sodium taurocholate into the bile-pancreatic duct, respectively. In addition, serum amylase and pancreatic tissue myeloperoxidase activities were measured to evaluate the pancreatic damage. According to gamma scintigraphy and biodistribution studies, accumulation times and distribution of (99m)Tc-MA and SA were different. While MA was highly uptake by reticuloendothelial system, SA was mostly excreted by kidneys and bladder. Compared with the mild acute pancreatitis group, treatment with MA significantly decreased the serum amylase activity and pancreas myeloperoxidase activity. Furthermore, the protease inhibitor molecule aprotinin has therapeutic potential in acute pancreatitis. Finally, MA may be suggested as a promising alternative for treatment of acute pancreatitis.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
2-Propanol, JIS special grade, ≥99.5%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
2-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Sigma-Aldrich
2-Propanol, suitable for HPLC
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O