MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Detection by ELISA of C-terminal proBNP in plasma from cats with cardiomyopathy.
Detection by ELISA of C-terminal proBNP in plasma from cats with cardiomyopathy.
Veterinary journal (London, England : 1997) (2015-09-02)
Philip F Solter, Mark A Oyama, Maggie C Machen, Dennis J Trafny, D David Sisson
ABSTRACT
The B-type natriuretic peptide prohormone (proBNP) is enzymatically cleaved into an inactive N-terminal peptide and a biologically active C-terminal peptide with many beneficial cardiorenal effects. The purpose of this study was to develop and test in cats with cardiomyopathy an immunoassay to quantify the concentrations of C-terminal proBNP in feline plasma. An anti-canine proBNP monoclonal antibody (UI-1021) was shown to have adequate binding affinity to proBNP 80-106 for use in a solid-phase immunoassay, and by epitope mapping to bind within positions 84-87 of feline proBNP. UI-1021 was paired with an affinity-purified rabbit polyclonal detection antibody to feline proBNP 100-106, in a sandwich ELISA with feline proBNP 80-106 standard. The linearity and analytical range and sensitivity of the assay were confirmed from 1.4 to 85 pmol/L. Spike recovery averaged 106.5% (95% confidence interval 78-135%). Within run and intra-assay coefficients of variation were <12%. A protease inhibitor mixture preserved proBNP 80-106 immunoreactivity for at least 5 days in plasma. Clinical verification of the ELISA was done using plasma from 13 cats with cardiomyopathy, whose C-terminal proBNP concentrations ranged from 1.7 to 78.8 pmol/L vs. <1.4-1.8 pmol/L in plasma from 18 healthy cats. Concentrations were found to be substantially lower than reported N-terminal proBNP concentrations, and similar to those of human heart failure patients where relative C-terminal BNP deficiencies have been proposed as contributory to the progression of the disease.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)