MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Quantification of nerolidol in mouse plasma using gas chromatography-mass spectrometry.
Quantification of nerolidol in mouse plasma using gas chromatography-mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2015-04-17)
Alexandre Yukio Saito, Rodrigo Antonio Ceschini Sussmann, Emilia Akemi Kimura, Maria Belen Cassera, Alejandro Miguel Katzin
ABSTRACT
Nerolidol is a naturally occurring sesquiterpene found in the essential oils of many types of flowers and plants. It is frequently used in cosmetics, as a food flavoring agent, and in cleaning products. In addition, nerolidol is used as a skin penetration enhancer for transdermal delivery of therapeutic drugs. However, nerolidol is hemolytic at low concentrations. A simple and fast GC-MS method was developed for preliminary quantification and assessment of biological interferences of nerolidol in mouse plasma after oral dosing. Calibration curves were linear in the concentration range of 0.010-5 μg/mL nerolidol in mouse plasma with correlation coefficients (r) greater than 0.99. Limits of detection and quantification were 0.0017 and 0.0035 μg/mL, respectively. The optimized method was successfully applied to the quantification of nerolidol in mouse plasma.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, suitable for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, suitable for residue analysis
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Hexane, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC