MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Semi solid matrix formulations of meloxicam and tenoxicam: an in vitro and in vivo evaluation.
Semi solid matrix formulations of meloxicam and tenoxicam: an in vitro and in vivo evaluation.
Archives of pharmacal research (2014-04-23)
Saritha Alladi, Nalini R Shastri
ABSTRACT
The objective of this study was to improve the dissolution and subsequently the therapeutic efficacy of poorly water soluble BCS class-II drugs meloxicam and tenoxicam, by lipid semi solid matrix (SSM) systems filled in hard gelatin capsules by liquid fill technology. The present research involved preparation of SSM formulations using Gelucire 44/14 as a carrier due to its self emulsifying, wetting and hydrophilic properties. The SSM capsules were characterized by assay, in vitro dissolution studies, moisture uptake, FTIR and DSC. The optimized formulations were also evaluated for their in vivo anti inflammatory activity in rat model. Six to ten fold enhancement in vitro drug release, in both acidic and basic media, was obtained with formulations containing drug to carrier in 1:6 ratio. The absence of drug peak in DSC scans indicated complete dissolution of the drug in carrier, while IR revealed no chemical interaction of pure drug and Gelucire 44/14. The optimized SSM formulations of meloxicam and tenoxicam showed a rapid decrease in paw edema with a significant increase in anti-inflammatory activity. The SSM formulations were successful in providing rapid release of drugs with improved dissolution and in vivo anti-inflammatory activity by liquid fill technology in hard gelatin capsules.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology