MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Mechanics of Viral Chromatin Reveals the Pressurization of Human Adenovirus.
Mechanics of Viral Chromatin Reveals the Pressurization of Human Adenovirus.
ACS nano (2015-10-23)
Alvaro Ortega-Esteban, Gabriela N Condezo, Ana J Pérez-Berná, Miguel Chillón, S Jane Flint, David Reguera, Carmen San Martín, Pedro J de Pablo
ABSTRACT
Tight confinement of naked genomes within some viruses results in high internal pressure that facilitates their translocation into the host. Adenovirus, however, encodes histone-like proteins that associate with its genome resulting in a confined DNA-protein condensate (core). Cleavage of these proteins during maturation decreases core condensation and primes the virion for proper uncoating via unidentified mechanisms. Here we open individual, mature and immature adenovirus cages to directly probe the mechanics of their chromatin-like cores. We find that immature cores are more rigid than the mature ones, unveiling a mechanical signature of their condensation level. Conversely, intact mature particles demonstrate more rigidity than immature or empty ones. DNA-condensing polyamines revert the mechanics of mature capsid and cores to near-immature values. The combination of these experiments reveals the pressurization of adenovirus particles induced by maturation. We estimate a pressure of ∼30 atm by continuous elasticity, which is corroborated by modeling the adenovirus mini-chromosome as a confined compact polymer. We propose this pressurization as a mechanism that facilitates initiating the stepwise disassembly of the mature particle, enabling its escape from the endosome and final genome release at the nuclear pore.
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%