MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Nanopatterned polystyrene-b-poly(acrylic acid) surfaces to modulate cell-material interaction.
Nanopatterned polystyrene-b-poly(acrylic acid) surfaces to modulate cell-material interaction.
Materials science & engineering. C, Materials for biological applications (2017-04-19)
Erlantz Lizundia, Míriam Sáenz-Pérez, David Patrocinio, Iskander Aurrekoetxea, Maria dM Vivanco, José Luis Vilas
ABSTRACT
In this work we explore the effect of surface nanoarchitecture of polystyrene (PS) and polystyrene-b-poly(acrylic acid) (PS-b-PAA) diblock copolymer films on cell viability. PS and PS-b-PAA have been nanopatterned at temperatures of 110, 120 and 140°C using nanoporous aluminium oxide membranes (AAO) as a template. Surface architecture strongly depends on the infiltration temperature and the nature of the infiltrated polymer. High patterning temperatures yield hollow fibre shape architecture at the nanoscale level, which substantially modifies the surface hydrophobicity of the resulting materials. Up to date very scarce reports could be found in the literature dealing with the interaction of microstructured/nanostructured polymeric surfaces with cancer cells. Therefore, MCF-7 breast cancer cells have been selected as a model to conduct cell viability assays. The findings reveal that the fine-tuning of the surface nanoarchitecture contributes to the modification of its biocompatibility. Overall, this study highlights the potential of AAO membranes to obtain well-defined tailored morphologies at nanoscale level and its importance to develop novel soft functional surfaces to be used in the biomedical field.
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%
Sigma-Aldrich
tert-Butyl acrylate, contains 10-20 ppm monomethyl ether hydroquinone as inhibitor, 98%
Sigma-Aldrich
Triton X-100, laboratory grade