MilliporeSigma
  • SHORT syndrome due to a novel de novo mutation in PRKCE (Protein Kinase Cɛ) impairing TORC2-dependent AKT activation.
SHORT syndrome due to a novel de novo mutation in PRKCE (Protein Kinase Cɛ) impairing TORC2-dependent AKT activation.
Human molecular genetics (2017-09-22)
Diana Alcantara, Frances Elmslie, Martine Tetreault, Eric Bareke, Taila Hartley, Jacek Majewski, Kym Boycott, A Micheil Innes, David A Dyment, Mark O'Driscoll
ABSTRACT
SHORT syndrome is a rare, recognizable syndrome resulting from heterozygous mutations in PIK3R1 encoding a regulatory subunit of phosphoinositide-3-kinase (PI3K). The condition is characterized by short stature, intrauterine growth restriction, lipoatrophy and a facial gestalt involving a triangular face, deep set eyes, low hanging columella and small chin. PIK3R1 mutations in SHORT syndrome result in reduced signaling through the PI3K-AKT-mTOR pathway. We performed whole exome sequencing for an individual with clinical features of SHORT syndrome but negative for PIK3R1 mutation and her parents. A rare de novo variant in PRKCE was identified. The gene encodes PKCε and, as such, the AKT-mTOR pathway function was assessed using phospho-specific antibodies with patient lymphoblasts and following ectopic expression of the mutant in HEK293 cells. Kinase analysis showed that the variant resulted in a partial loss-of-function. Whilst interaction with PDK1 and the mTORC2 complex component SIN1 was preserved in the mutant PKCε, it bound to SIN1 with a higher affinity than wild-type PKCε and the dynamics of mTORC2-dependent priming of mutant PKCε was altered. Further, mutant PKCε caused impaired mTORC2-dependent pAKT-S473 following rapamycin treatment. Reduced pFOXO1-S256 and pS6-S240/244 levels were also observed in the patient LCLs. To date, mutations in PIK3R1 causing impaired PI3K-dependent AKT activation are the only known cause of SHORT syndrome. We identify a SHORT syndrome child with a novel partial loss-of-function defect in PKCε. This variant causes impaired AKT activation via compromised mTORC2 complex function.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder