Merck
20479
GlasSeal Capillary Column Connector, Borosilicate Glass
pkg of 12 ea
登入 查看組織和合約定價
選擇尺寸
暫時無法取得訂價和供貨情況
關於此項目
分類程式碼代碼:
41121800
跳轉至
技術服務
需要援助?我們經驗豐富的科學家團隊隨時樂意為您提供支援。
讓我們為您提供幫助
比較類似項目
查看完整比較
Show Differences
1 of 4
本產品
236272362823632
packaging
pkg of 12 ea
packaging
pkg of 5 ea
packaging
pkg of 25 ea
packaging
pkg of 3 ea
technique(s)
gas chromatography (GC): suitable
technique(s)
gas chromatography (GC): suitable
technique(s)
gas chromatography (GC): suitable
technique(s)
gas chromatography (GC): suitable
一般說明
Column connectors are useful for attaching a guard column/retention gap to an analytical column, or for repairing a broken column.
Quickly connect two pieces of fused silica tubing of the same or different diameters without the need for tools. The leak-free connection is useful to connect a guard column or transfer line, repair a broken column, or connect two columns (same or different phases). Silanized for an inert inside surface. For use with 0.20 - 0.53 mm I.D. fused silica tubing.
其他說明
We offer a variety of chromatography accessories including analytical syringes
法律資訊
從最近期的版本中選擇一個:
分析證明 (COA)