Skip to Content
MilliporeSigma

Skip To

GF20040286

Polyaramid

(polymetaphenylene isophthalamide), film, thickness 0.05 mm, L 2 m, type t410

Synonym(s):

AR311050

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$810.00

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Manufacturer/tradename:
Goodfellow 200-402-86

$810.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

film

manufacturer/tradename

Goodfellow 200-402-86

L

2 m

thickness

0.05 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF48960322GF78394067GF97576071
description

(polymetaphenylene isophthalamide), film, thickness 0.05 mm, L 2 m, type t410

description

(polymetaphenylene isophthalamide), film, thickness 0.05 mm, L 1 m, type t410

description

(polymetaphenylene isophthalamide), film, thickness 0.51 mm, size 200 × 304 mm, type t410

description

(polymetaphenylene isophthalamide), film, thickness 0.76 mm, L 2 m, coil width 63.5 mm

manufacturer/tradename

Goodfellow 200-402-86

manufacturer/tradename

Goodfellow 489-603-22

manufacturer/tradename

Goodfellow 783-940-67

manufacturer/tradename

Goodfellow 975-760-71

L

2 m

L

1 m

L

-

L

2 m

form

film

form

film

form

film

form

film

thickness

0.05 mm

thickness

0.05 mm

thickness

0.51 mm

thickness

0.76 mm


Still not finding the right product?

Explore all of our products under Polyaramid


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters