MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Two leucines in the N-terminal MAPK-docking site of tomato SlMKK2 are critical for interaction with a downstream MAPK to elicit programmed cell death associated with plant immunity.
Two leucines in the N-terminal MAPK-docking site of tomato SlMKK2 are critical for interaction with a downstream MAPK to elicit programmed cell death associated with plant immunity.
FEBS letters (2013-04-16)
Chang-Sik Oh, JeeNa Hwang, Min-Seon Choi, Byoung-Cheorl Kang, Gregory B Martin
ABSTRACT
Tomato MAPK kinase SlMKK2 is a key protein regulating immunity-associated programmed cell death (PCD) in plants. We examined the role of the N-terminal MAPK-docking site (or D-site) of SlMKK2 in PCD elicitation. In vivo assays revealed that SlMKK2 interacted with the downstream MAPK SlMPK3 independent of PCD elicitation and two conserved leucines in the D-site were required for both interaction with SlMPK3 and PCD elicitation. These results demonstrate that two leucines in the D-site of SlMKK2 play a critical role in regulation of signal transfer to the downstream MAPK by regulating their physical interaction.
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, 99%, FG
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland