MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Resveratrol possesses protective effects in a pristane-induced lupus mouse model.
Resveratrol possesses protective effects in a pristane-induced lupus mouse model.
PloS one (2014-12-17)
Zhuo-Long Wang, Xiao-Fang Luo, Meng-Tao Li, Dong Xu, Shuang Zhou, Hou-Zao Chen, Na Gao, Zhen Chen, Ling-Ling Zhang, Xiao-Feng Zeng
ABSTRACT
Systemic lupus erythematosus (SLE) is a multisystemic autoimmune disease characterized by the production of autoantibodies. To date, no therapy has been found to satisfactorily treat SLE. SIRT1 deficiency results in the development of an autoimmune syndrome in mice, including a high titer of anti-nuclear antibody in serum, immunoglobulin deposition in the kidney, and immune complex glomerulonephritis. Resveratrol is an activator of SIRT1 and possesses anti-inflammation and immune-regulatory properties. To evaluate the preventative effects of resveratrol on a pristane-induced lupus animal model and assess its putative immune modulation effects. BALB/c mice received a single intraperitoneal injection of 0.5 ml of pristane on day 1 and then various doses of resveratrol were given to the mice daily starting on day 2 and continuing for seven months. The autoantibodies in serum and supernatants were measured. Single cells isolated from spleen, isolated CD4+ T cells, and CD19+ B cells were cultured with or without resveratrol in vitro and assessed by flow cytometry. Resveratrol attenuated proteinuria, immunoglobuin depositon in kidney, and glomerulonephritis as well as IgG1 and IgG2a in serum in pristane-induced lupus mice. Resveratrol also suppressed CD69 and CD71 expression on CD4+ T cells as well as CD4+ T cell proliferation, induced CD4+ T cell apoptosis, and decreased CD4 IFNγ+ Th1 cells and the ratio of Th1/Th2 cells in vitro. In vitro antibody production and proliferation of B cells were also inhibited. Resveratrol possesses protective effects in pristane-induced lupus mice and may represent a novel approach for the management of SLE.
Product Description
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Resveratrol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Pristane, synthetic, ≥98% (GC)
SAFC
Pristane, synthetic, liquid, sterile-filtered, BioReagent