MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Flavobacterium vireti sp. nov., isolated from soil.
Antonie van Leeuwenhoek (2015-03-31)
Hina Singh, Juan Du, KyungHwa Won, Jung-Eun Yang, Shahina Akter, Ki-Young Kim, Tae-Hoo Yi
ABSTRACT
A novel Gram-negative, aerobic, yellow-pigmented, non-motile and rod-shaped bacterium, designated as THG-SM1(T), was isolated from field soil collected from Suwon, South Korea. The strain was found to grow optimally at 28 °C, at pH 7.0 and in the absence of NaCl. Based on 16S rRNA gene sequence similarities, strain THG-SM1(T) belongs to the genus Flavobacterium and is most closely related to Flavobacterium terrae KACC 11731(T), followed by Flavobacterium columnare KACC 11683(T) and Flavobacterium enshiense KCTC 23775(T). The DNA G+C content of the novel isolate was determined to be 38.5 mol%. In DNA-DNA hybridization tests, the DNA relatedness between strain THG-SM1(T) and its closest phylogenetic neighbour F. terrae was below 50 %. Flexirubin-type pigments were found to be present. The major polar lipid and isoprenoid quinone were phosphatidylethanolamine and menaquinone 6 (MK-6), respectively. The main cellular fatty acids were identified as iso-C15:1G, iso-C15:0 3OH, iso-C16:0 and iso-C15:0. The DNA-DNA hybridization result and differentiating chemotaxonomic and phenotypic characteristics showed that strain THG-SM1(T) represents a novel species of the genus Flavobacterium, for which the name Flavobacterium vireti sp. nov. is proposed. The type strain is THG-SM1(T) (=KACC 18371(T) = CCTCC AB2014312(T)).
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from Escherichia coli strain B, Genomic, unsheared
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Ninhydrin, SAJ special grade, ≥98.0%
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Iron(III) citrate, technical grade
Supelco
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis
Sigma-Aldrich
p-Phenylenediamine, ≥99.0% (GC/NT)
Sigma-Aldrich
p-Phenylenediamine, 98% (GC)