MilliporeSigma
Antibody Drug Conjugate Manufacturing
Illustration depicting antibody-drug conjugate (ADC) used to deliver highly active pharmaceutical ingredients
Antibody-drug conjugates (ADCs) are therapeutics that use monoclonal antibodies (mAbs) to deliver highly potent active pharmaceutical ingredient (HPAPI) to target cells. ADCs comprise a monoclonal antibody (mAb) conjugated to a payload, via a stable linker. The linker ensures that the payload remains attached to the antibody until it reaches the target site, thus minimizing off-target toxicity. Although most approved ADCs, and those in development, target various cancer indications, these complex therapeutics hold significant promise for treating a variety of other diseases.
Due to the complexities of ADC manufacturing, outsourcing to an experienced CDMO can be a strategic decision to navigate the challenges of ADC production, reduce risks, and accelerate the timeline to clinic and to market.
Whether you are considering outsourcing to a CDMO or seeking to enhance your in-house capabilities with high-quality raw materials and tailored equipment, we are here to support you at every stage of your process. Contact us to learn more about our comprehensive solutions designed to meet your ADC manufacturing needs.
Challenges of ADC development and manufacturing
The development and manufacturing of ADCs is more complex than that of mAbs, requiring significant technical expertise and robust containment measures to maintain operator safety. Typically, expertise in HPAPI and large molecule development, linker chemistry, and conjugation is needed, combined with analytical capabilities to characterize and demonstrate purity, homogeneity, and stability of the ADC product. In addition, manufacturing suites usually contain specialized equipment designed to mitigate the risk of operator exposure to the highly cytotoxic payloads.
Successful development and consistent production of the desired ADC molecule depends on several factors including:
  • Conjugation: Must not affect mAb functionality; can be non-site-specific or site-specific, dependent on the method of conjugation.
  • Linker Design: Affects the ADC stability following administration to the patient, efficiency of payload release at the target site, pharmacokinetics, efficacy, and toxicity.
  • Drug-to-Antibody Ratio (DAR): The desired average number of HPAPI or drug molecules conjugated to the mAb must be achieved. A low DAR can reduce effectiveness of the ADC while a high DAR can negatively impact the mAb structure, stability, target binding, and overall safety profile.
  • Efficient Purification: Optimized to reliably remove process related contaminants, residual linkers and cytotoxic HPAPI payload.
  • Consistent Purification: To maintain the desired therapeutic effect, for any given DAR there should be low variability in the ADC composition.
  • Reliable Containment: Is critical throughout the manufacturing workflow to maintain operator safety. Single-use systems provide a flexible, safe, and reliable solution for successful ADC manufacturing.
  • Stable and Safe Formulation: Parenteral administration requires the selection of high-quality excipients. ADC stability, solubilization, and administrability are required for optimal therapeutic efficacy.
Throughout the ADC manufacturing process, it is critical to select products and technologies that ensure high operator safety, reduce contamination risk, and support seamless scalability from development to GMP production. Single-use systems provide a safe, reproducible, and scalable solution, supporting high yields that meet target quality attributes. Comprehensive documentation on extractables and leachables from the supplier, facilitates risk assessment, and accelerates your path to successful and safe ADC manufacturing.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents
Workflow
mab Preparation
mAb Preparation
Preparation of the mAb involves possible modifications through chemical or enzymatic reactions. At this stage, preliminary conjugation of the mAb to the linker can be performed, requiring careful monitoring and control of the reaction.
Learn more about our advanced solutions for mAb development and manufacturing: Monoclonal Antibodies (mAbs)
    Conjugation
    The HPAPI payload or payload-linker complex reagents are usually prepared separately. Successful conjugation optimizes reagent addition, ensures proper mixing with monitoring and control of the conjugation reaction. 
    Learn more about ADC manufacturing: Conjugation
      Purification
      Chromatography and/or tangential flow filtration (TFF) is used to purify the ADC. The goal is to remove unwanted species, such as molecules with an undesirable DAR, aggregates, residual unconjugated linker-payload, and reaction solvents. Conjugated payloads can promote aggregation of the ADC, especially if very hydrophobic, presenting an additional challenge for purification.
      Further Purification and Final Formulation
      Additional purification may be necessary to reduce contaminants and exchange the ADC to formulation buffer. Maintaining ADC solubility is crucial, especially given the typically high hydrophobicity. The use of high-quality chemicals such as buffers, pH adjusters, surfactants, stabilizers, and viscosity-reducing excipients supports efficient purification and a stable final formulation.
      Sterile Filtration
      The formulated bulk ADC is sterile filtered prior to filling or shipment for filling offsite. Proper containment is critical to maintain sterility of the ADC bulk and assure operator safety from the highly toxic HPAPI payload.
      Fill Finish
      The bulk ADC drug product is sterile filtered prior to filling. Well-designed final filtration and filling assemblies can minimize product loss, maximize ADC yield while enabling filter integrity testing and maintaining sterility and the containment needed for operator safety.
      Featured Categories
      A sleek, modern Mobius mixer used in pharmaceutical and biopharma manufacturing. The device features a cylindrical design with a digital control panel, ensuring precise mixing and processing of materials.
      Single-Use Mixers
      Our complete range of Mobius® single-use mixing solutions can meet your needs for mixing pharmaceutical ingredients from intermediate to final drug products, as well as preparation of process solutions such as buffers and media.
      Shop Products
      The Mobius® FlexReady Solution with Smart Flexware™ Assemblies is a flexible system, allowing end user to have a single, scalable system for chromatography and tangential flow filtration at various scales
      Ultrafiltration & Diafiltration
      Ultrafiltration & diafiltration are important steps throughout bioprocessing to increase capacity, concentration, and recovery.
      Shop Products
      Vantage® columns
      Ion Exchange Chromatography Resins
      Ion exchange (IEX) chromatography separates biomolecules based on difference in charge...
      Shop Products
      Liquid Formulation
      Biopharmaceutical Formulation
      Biologics are therapeutics that can be based on proteins, peptides, viruses, viral vectors, or nucleic acids. They require a defined environment to remain intact and functional throughout the manufacturing and formulation process, as well as during long-term storage.
      Shop Products