[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Bioelectronics](https://www.sigmaaldrich.com/US/en/applications/materials-science-and-engineering/bioelectronics)Nanomaterials for Biomedical Applications
# Nanomaterials for Biomedical Applications
Dendrimers are a class of nano-sized materials that are used to develop life-saving technologies. Single-wall carbon nanotubes are used to make miniaturized sensors for DNA, pathogens, and chemicals – for example glucose sensors. Multi-wall nanotubes, nanoclays, and ceramic nanoparticles are incorporated in ultra-strong polymer nanocomposites that will be used in orthopedic, for example joint-replacement, applications. Nanoparticles can also be used to make nano-textured substrates to support attachment and growth of cells for tissue engineering. Hydroxyapatite nanopowders (Product No. [677418](https://www.sigmaaldrich.com/US/en/product/aldrich/677418)), incorporated in biodegradable polymer composites1 or deposited on biocompatible substrates2, have been shown to promote adhesion and proliferation of bone-forming (osteoblast) cells. We are looking forward to offering new nanomaterials tools to accelerate research in biotechnology and medicine.
[](https://www.sigmaaldrich.com/catalog/product/aldrich/677418)
__Figure 1:__ Hydroxyapatite Nanopowder
- Molecular Formula \[Ca5(OH)(PO4)3]x
- Average Particle Size 150 – 200 nm
- Surface Area > 9.4 m2/g
- Available in 5 g pack size
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Kim S, Sun Park M, Jeon O, Yong Choi C, Kim B. 2006. Poly(lactide-co-glycolide)/hydroxyapatite composite scaffolds for bone tissue engineering. Biomaterials. 27(8):1399-1409. [https://doi.org/10.1016/j.biomaterials.2005.08.016](https://doi.org/10.1016/j.biomaterials.2005.08.016)
2\.
Sato M, Sambito MA, Aslani A, Kalkhoran NM, Slamovich EB, Webster TJ. 2006. Increased osteoblast functions on undoped and yttrium-doped nanocrystalline hydroxyapatite coatings on titanium. Biomaterials. 27(11):2358-2369. [https://doi.org/10.1016/j.biomaterials.2005.10.041](https://doi.org/10.1016/j.biomaterials.2005.10.041)
__Related Articles__
- [Graphene Inks for Electronics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/graphene-inks-for-printed-electronics)
- [Functionalized Graphene Oxide: Properties & Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/functionalized-graphene-oxide)
- [2D Materials in Biosensing](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/2d-biosensing)
- [Conducting Polymer Device Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/applications-of-conducting-polymer-devices-in-life-sciences)
- [Conducting Bio-based Polymers and Composites for Advanced Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/conducting-bio-based-polymers-and-composites)
- [Conductive Polymers for Advanced Micro- and Nano-fabrication Processes](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/conductive-polymers)
- [Graphene in Biotechnology](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/graphene-in-biotechnology)
- [Nano Minerals: Nanoclays](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/nano-minerals-nanoclays)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
An unknown error has occured.