Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

MilliporeSigma

Skip To

GF00712437

Copper

foil, thickness 0.1 mm, length 5 m, coil width 30 mm

Synonym(s):

CU000558

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$650.00

About This Item

Linear Formula:
Cu
CAS Number:
Molecular Weight:
63.55
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141711
MDL number:
eCl@ss:
38150101

$650.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

foil

manufacturer/tradename

Goodfellow 007-124-37

resistivity

1.673 μΩ-cm, 20°C

L

5 m

coil width

30 mm

thickness

0.1 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF51053699GF86241719GF94916188
description

foil, thickness 0.1 mm, length 5 m, coil width 30 mm

description

tube, 100mm, outside diameter 3.0mm, inside diameter 1.2mm, wall thickness 0.9mm, as drawn, 99.9%

description

foil, thickness 0.0015 mm, purity 99.97%, size 25 × 25 mm

description

foil, thickness 0.05 mm, length 50 m, coil width 12 mm

form

foil

form

tubes

form

foil

form

foil

manufacturer/tradename

Goodfellow 007-124-37

manufacturer/tradename

Goodfellow 510-536-99

manufacturer/tradename

Goodfellow 862-417-19

manufacturer/tradename

Goodfellow 949-161-88

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters