Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

MilliporeSigma

Skip To

GF04550880

Polyamide - Nylon 6

(PA 6), sheet, thickness 10 mm, size 500 × 500 mm

Synonym(s):

AM303100

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$810.00

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100

$810.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

sheet

manufacturer/tradename

Goodfellow 045-508-80

size

500 × 500 mm

thickness

10 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF12836404GF98123520GF65440841
description

(PA 6), sheet, thickness 10 mm, size 500 × 500 mm

description

(PA 6), sheet, thickness 10 mm, size 300 × 300 mm

description

(PA 6), sheet, thickness 10 mm, size 150 × 150 mm, condition oil-filled nylon ′6′

description

(PA 6), sheet, thickness 10 mm, size 300 × 300 mm, condition oil-filled nylon ′6′

manufacturer/tradename

Goodfellow 045-508-80

manufacturer/tradename

Goodfellow 128-364-04

manufacturer/tradename

Goodfellow 981-235-20

manufacturer/tradename

Goodfellow 654-408-41

thickness

10 mm

thickness

10 mm

thickness

10 mm

thickness

10 mm

form

sheet

form

sheet

form

sheet

form

sheet

size

500 × 500 mm

size

300 × 300 mm

size

150 × 150 mm

size

300 × 300 mm


Still not finding the right product?

Explore all of our products under Polyamide - Nylon 6


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters