Skip to Content
MilliporeSigma

Skip To

GF33720229

Tetrafluoroethylene-Perfluoro(alkoxy vinyl ether)

(alkoxy vinyl ether), (pfa), film, thickness 0.052 mm, L 10 m

Synonym(s):

FV321050

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$1,360.00

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Manufacturer/tradename:
Goodfellow 337-202-29

$1,360.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

film

manufacturer/tradename

Goodfellow 337-202-29

L

10 m

thickness

0.052 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF10317248GF37723844GF53253389
description

(alkoxy vinyl ether), (pfa), film, thickness 0.052 mm, L 10 m

description

(alkoxy vinyl ether), (pfa), film, thickness 0.052 mm, L 0.5 m

description

(alkoxy vinyl ether), (pfa), film, thickness 0.052 mm, L 5 m

description

(alkoxy vinyl ether), (pfa), film, thickness 0.052 mm, size 150 × 150 mm

manufacturer/tradename

Goodfellow 337-202-29

manufacturer/tradename

Goodfellow 103-172-48

manufacturer/tradename

Goodfellow 377-238-44

manufacturer/tradename

Goodfellow 532-533-89

L

10 m

L

0.5 m

L

5 m

L

-

form

film

form

film

form

film

form

film

thickness

0.052 mm

thickness

0.052 mm

thickness

0.052 mm

thickness

0.052 mm


Still not finding the right product?


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters