Skip to Content
MilliporeSigma

Skip To

GF45889396

Aluminum

foil, thickness 0.25 mm, length 1 m, purity 99%

Synonym(s):

AL000622

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$435.00

About This Item

Linear Formula:
Al
CAS Number:
Molecular Weight:
26.98
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141702
MDL number:
eCl@ss:
38120301

$435.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

form

foil

autoignition temp.

1400 °F

manufacturer/tradename

Goodfellow 458-893-96

resistivity

2.6548 μΩ-cm

L

1 m

thickness

0.25 mm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF25185977GF27935950GF85240694
description

foil, thickness 0.25 mm, length 1 m, purity 99%

description

foil, thickness 2.0 mm, size 150 × 150 mm, high purity 99.999%

description

foil, thickness 0.3 mm, length 2 m, purity 99%

description

honeycomb, 600x600mm, thickness 20mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%

form

foil

form

foil

form

foil

form

honeycomb

assay

99%

assay

99.999%

assay

99%

assay

99.5%

manufacturer/tradename

Goodfellow 458-893-96

manufacturer/tradename

Goodfellow 251-859-77

manufacturer/tradename

Goodfellow 279-359-50

manufacturer/tradename

Goodfellow 852-406-94

mp

660.37 °C (lit.)

mp

660.37 °C (lit.)

mp

660.37 °C (lit.)

mp

660.37 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters