Skip to Content
MilliporeSigma

Skip To

GF46098546

Polybutylene Terephthalate - 30% Glass Fiber Reinforced

(PBT 30% GFR), rod, diam. 20 mm, L 1000 mm, condition machining feedstock

Synonym(s):

ES357920

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$628.00

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100

$628.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

rod

manufacturer/tradename

Goodfellow 460-985-46

L

1000 mm

diam.

20 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF20463757GF17214005GF31363714
description

(PBT 30% GFR), rod, diam. 20 mm, L 1000 mm, condition machining feedstock

description

(PBT 30% GFR), rod, diam. 20 mm, L 200 mm, condition machining feedstock

description

(PBT 30% GFR), rod, diam. 40 mm, L 200 mm, condition machining feedstock

description

(PBT 30% GFR), rod, diam. 20 mm, L 100 mm, condition machining feedstock

diam.

20 mm

diam.

20 mm

diam.

40 mm

diam.

20 mm

L

1000 mm

L

200 mm

L

200 mm

L

100 mm

manufacturer/tradename

Goodfellow 460-985-46

manufacturer/tradename

Goodfellow 204-637-57

manufacturer/tradename

Goodfellow 172-140-05

manufacturer/tradename

Goodfellow 313-637-14

form

rod

form

rod

form

rod

form

rod

polybutylene terephthalate GF30 for 3D printing

high strength polybutylene terephthalate with glass fibers, polybutylene terephthalate glass fiber composite for 3D printing, polybutylene terephthalate with glass fiber reinforcement, 30% glass fiber reinforced polybutylene terephthalate, glass fiber reinforced polybutylene terephthalate resin, industrial grade polybutylene terephthalate with 30% GF, 30% GF polybutylene terephthalate for injection molding, PBT 30% glass fiber reinforced for automotive applications, PBT GF30 engineering plastic, polybutylene terephthalate glass fiber reinforced sheets, PBT GF30 stock for manufacturing, polybutylene terephthalate composites for electronics, PBT 30% glass fiber reinforced plastic, reinforced PBT material for industrial applications

polybutylene terephthalate resin 30% glass fiber, engineering plastic polybutylene terephthalate 30% GF, high-strength polybutylene terephthalate for manufacturing, polybutylene terephthalate glass fiber reinforced properties, PBT GF30 material for injection molding, polybutylene terephthalate 30% glass fiber composite, industrial grade polybutylene terephthalate 30% GF, polybutylene terephthalate for industrial use, polybutylene terephthalate composite with glass fiber, polybutylene terephthalate technical data sheet, 30% glass fiber reinforced polybutylene terephthalate, PBT with 30% glass fiber reinforcement, 30% glass fiber PBT for automotive applications, PBT 30% GF for electrical insulation applications

-


Still not finding the right product?


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters