Skip to Content
MilliporeSigma

Skip To

GF25762136

Polypropylene

(PP), film, thickness 0.04 mm, L 20 m, condition biaxially oriented

Synonym(s):

PP301341

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$1,050.00

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Manufacturer/tradename:
Goodfellow 257-621-36

$1,050.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

film

manufacturer/tradename

Goodfellow 257-621-36

L

20 m

thickness

0.04 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF25130326GF22038724GF18538678
description

(PP), film, thickness 0.04 mm, L 20 m, condition biaxially oriented

description

(PP), film, thickness 0.04 mm, L 10 m, condition biaxially oriented

description

(PP), film, thickness 0.04 mm, L 1 m, condition biaxially oriented

description

(PP), film, thickness 0.04 mm, L 0.5 m, condition biaxially oriented

manufacturer/tradename

Goodfellow 257-621-36

manufacturer/tradename

Goodfellow 251-303-26

manufacturer/tradename

Goodfellow 220-387-24

manufacturer/tradename

Goodfellow 185-386-78

L

20 m

L

10 m

L

1 m

L

0.5 m

form

film

form

film

form

film

form

film

thickness

0.04 mm

thickness

0.04 mm

thickness

0.04 mm

thickness

0.04 mm


Still not finding the right product?

Explore all of our products under Polypropylene


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters